Learn More
MTC Bio Western Blot Boxes, Removable Lid, Opaque Black, for Novex Minigel, 8.6 x 8.6 x 2.8cm, 10/pk
Supplier: MTC Bio B13008BK
This item has a minimum qty of 2 per supplier requirements.
No matter what size your Western blot is, theres a box that fits it perfectly. With boxes that are correctly matched to common blot sizes, you can fit more blots on shakers/rockers while keeping reagent and antibody usage to an absolute minimum. Additionally, all corners are formed at 90 angles and the bottom surface is uniformly flat, further reducing consumption of costly reagents and antibodies. (For example, popular mini-gels require only about 4ml.) Many boxes are also available in light-blocking opaque black for light sensitive samples
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.