Learn More
Andwin Scientific Block Filing Cabinet No. 4192
Supplier: Andwin Scientific 6251501
The Tissue-Tek Lab Aid block filing cabinet by Electron Microscopy Sciences provides secure storage and organization of blocks that have been embedded with the use of tissue cassettes, biopsy cassettes, mesh cassettes, mega cassettes, processing or embedding cassettes, and emberring rings. Its one-piece design and lack of hinges and doors allow dust-free storage. There are six drawers that are impact-resistant. This product is very compact in size, measuring 17" x 9" x 15"
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.