Learn More
Andwin Scientific BLADES 50/PK (10PKS/CS)
Supplier: Andwin Scientific 25608964
The Tissue-Tek� Accu-Edge� low-profile blades are sharp and disposable. These blades can section specimens without striations, distortion, or chattering. They are compatible with all Accu-Edge� blade holders for microtomy and cryotomy.