Learn More
Andwin Scientific BLADES 5/CS
Supplier: Andwin Scientific MDT4796
The Tissue-Tek� Accu-Edge� replaceable Sharp/Blunt blade is an ideal tool for performing anatomical sectioning of tissues. The blades can be easily inserted and removed, which removes the time, expense, and incovenience of sharpening. This product is autoclavable and is sold in a case of 5.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.