Learn More
Agilent Technologies BL21-CODON PLUS RIPL CELLS
Supplier: Agilent Technologies 230280
BL21-CodonPlus (DE3)-RIPL Competent Cells, 10 x 0.1mL, These cells contain extra copies of rare E. coli argU, ileY, leuW, proL tRNA genes which corrects for codon bias and dramatically improves expression of sequences from other organisms