Learn More
Enzo Life Sciences Bisindolylmaleimide III (1mg)
Supplier: Enzo Life Sciences ALX270051M001
PKC inhibitor. Potent and selective inhibitor of protein kinase C (PKC). Formula: C23H20N4O2. MW: 384.4. Purity: ≥98%. Solubility: Soluble in methanol or DMSO. Long Term Storage: -20°C. Handling: Protect from light.