Learn More
Enzo Life Sciences Bisindolylmaleimide II (1mg). CAS: 137592-45-1
SDP
Supplier:  Enzo Life Sciences ALX270050M001
Encompass_Preferred
PKC inhibitor. Formula: C27H26N4O2. MW: 438.5. Purity: ≥97% (HPLC). Appearance: Orange solid. Solubility: Soluble in DMSO or 100% ethanol. Long Term Storage: -20°C. Handling: Protect from light.
Catalog No. 50-200-8381
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.