Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(2-ethylhexyl) Phthalate Solution, 5000μg/mL in methanol, Ultra Scientific

Catalog No. USEPA1083
Encompass
Click to view available options
Quantity:
1 x 1 mL

Chemical Identifiers

CAS 117-81-7, 67-56-1
Molecular Formula C24H38O4
Molecular Weight (g/mol) 390.564
InChI Key BJQHLKABXJIVAM-UHFFFAOYSA-N
PubChem CID 8343
ChEBI CHEBI:17747
IUPAC Name bis(2-ethylhexyl) benzene-1,2-dicarboxylate
SMILES CCCCC(CC)COC(=O)C1=CC=CC=C1C(=O)OCC(CC)CCCC

Specifications

Quantity 1 x 1 mL
CAS Max % 99.36, 0.63
Concentration or Composition (by Analyte or Components) 5000μg/mL
Analyte or Component Names methanol,bis(2-chloroisopropyl) ether
Solvent or Matrix methanol
Chemical Name or Material Bis(2-ethylhexyl) Phthalate Solution
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.