Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals Biotin Sufficient Diet, Pellet

Biotin sufficient diet. Contains casein, fiber and oil.

$1031.00 - $3897.00

Specifications

Product Type Animal Diet
Autoclavable Autoclavable
Disposable Single-use
Format Pellet
Sterility Non-sterile
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MP296040810
View Documents
MP Biomedicals
0296040810
10 kg
Each for $1,031.00
Add to Cart
 
MP296040820
View Documents
MP Biomedicals
0296040820
20 kg
Each for $1,786.00
Add to Cart
 
MP296040850
View Documents
MP Biomedicals
0296040850
50 kg
Each for $3,897.00
Add to Cart
 
Description

Description

  • Biotin sufficient diet
  • Contains casein, fiber, oil, salt mixture and MP Bio vitamin diet fortification mixture
Specifications

Specifications

Animal Diet
Single-use
Non-sterile
1 ft.
Autoclavable
Pellet
Refrigerated
0.3048 m
Videos
SDS
Documents

Documents

Product Certifications

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.