Learn More
Electron Microscopy Sciences Biopsy Bags, Size: 74 x 94 mm., 100/PK
Supplier: Electron Microscopy Sciences 6232603
These bags are made from porous paper which provides maximum tissue safety during processing. They permit proper fluid exchange and drainage. To Use simply introduce the specimen into the bag and fold it enclosing the cassette or capsule. You are now ready to process as usual. Size: 74 x 94 mm.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.