Learn More
Enzo Life Sciences BIOARRAY™ Deoxyoligo B2 (150Reactions)
SDP
Supplier:  Enzo Life Sciences ENZ42662150
Encompass_Preferred
BIOARRAY™ Deoxyoligo B2 is a synthetic biotin-labeled sequence that can be used in eukaryotic expression assays and in aiding software programs such as the Affymetrix Microarray Suite by aligning the grid with the array image. The Control Deoxyoligo B2, together with the Eukaryotic Hybridization Controls (included in ENZ-42661-30 or ENZ-42661-150), provides an internal control for assaying quality and performance. This is done by adding the controls to biotin-labeled target cRNA before hybridization to probe sequences on the array. Deoxyoligo B2 is supplied as a 3 nM solution. Long Term Storage: -80°C.
Catalog No. 50-201-2529
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.