Learn More
Enzo Life Sciences BIO-PROBE Blur 8 probe (2µg)
Supplier: Enzo Life Sciences ENZ40849
The Blur 8 BIO-PROBE labeled probe is prepared by nick translation of a clone of the human alu repeat sequence. Fragment size range: 100-1000 base pairs (as estimated by agarose gel electrophoresis). Concentration: 20µg/ml. Formulation: Liquid. In 10mM TRIS HCl, pH 7.5, containing 1mM EDTA. Long Term Storage: -20°C.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.