Learn More
Enzo Life Sciences BIO-PROBE Adenovirus probe (2µg)
Supplier: Enzo Life Sciences ENZ40834
The Adenovirus BIO-PROBE labeled probe is prepared by nick translation of cloned fragments of the adenovirus 5 genome. The fragments constitute a total of 20 kb of DNA, approximately 56% of the genome. The probe is specific for adenovirus and, in addition to hybridization to adenovirus 5 sequences, the probe will cross hybridize with adenovirus types 4, 7, 11, 20, 40 and 41 due to their considerable sequence homology. Fragment size range: 100-1000 base pairs (as estimated by agarose gel electrophoresis). Concentration: 20µg/ml. Formulation: Liquid. In 10mM TRIS HCl, pH 7.5, containing 1mM EDTA. Long Term Storage: -20°C.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.