Learn More
Andwin Scientific BIO-HAZARD BAGS 23X19 200CS
SDP
Supplier:  Andwin Scientific JX1002F
Encompass_Preferred
The bio-hazard bags are autoclavable and resistant to temperatures up to 135 deg.C. This product can also withstand puncture, tears or leaks. The bags have a dimension of 19" x 23" and thickness of 2.0 mil. This is labeled with a biohazard symbol and use guidelines. It is sold in a case of 200.
Catalog No. NC9049925
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.