Learn More
Andwin Scientific BIO HAZ BAGS 8X12 W/IND 400CS
SDP
Supplier:  Andwin Scientific 14220046
Encompass
The 8x12 orange biohazard bag is ideal for processing biohazardous materials that require autoclaving prior to disposal. It is constructed from 2mil-thick rigid, high integrity polypropylene that can withstand 135C (275F) maximum autoclave temperature, resisting punctures, tears and leaks. It features a twist-tie closure that permits the bag to expand and vent steam during cooling. This model features an indicator statement Contents Have Been Autoclaved printed in a chemically active ink that darkens when autoclaving is complete, providing instant proof. It is sold in a case of 400.
Catalog No. NC9741695
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.