Learn More
Enzo Life Sciences Big Endothelin-1 (rat), ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ADI900073
Encompass_Preferred
The Big Endothelin-1 (rat), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results overnight + 1 hour. Alternative name: Big ET-1. Sensitivity: 0.72 pg/ml (range 0.78 - 100 pg/ml). UniProt ID: P22388. Assay Time: Overnight + 1 hour. Applications: ELISA, Colorimetric detection . Species reactivity: Rat . Content: Microtiter plate, Labeled antibody concentrate, Antibody diluent, Assay buffer, Wash buffer concentrate, Standard, TMB Substrate, Stop solution.
Catalog No. 50-200-6835
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.