Learn More
Enzo Life Sciences Betulinic acid (~95%) (500mg). CAS: 472-15-1
SDP
Supplier:  Enzo Life Sciences ALX350298M500
Encompass_Preferred
Antitumor and anti-HIV agent. Induces apoptosis by activating mitochondrial permeability transition (MPT). Inhibits NF-κB activation and NF-κB-regulated gene expression induced by carcinogens and inflammatory stimuli. Decreases expression of Bcl-2 and cyclin D1. Potent proteasome activator. Alternative name: 3beta-Hydroxy-20(29)-lupaene-28-oic acid. Purity: ≥90% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: +4°C.
Catalog No. 50-200-8836
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.