Learn More
Thermo Scientific™ BetaBasic™ 18 Guard Cartridges
Available in limited dimensions. Please contact Customer Service for more details.
$822.00
Specifications
Carbon Load | 13% |
---|---|
pH | 2 to 12 |
Length (Metric) | 10 mm |
Pore Size | 150Å |
Surface Area | 200 m2/g |
Catalog Number | Mfr. No. | Diameter (Metric) | Length (Metric) | Particle Size | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (Metric) | Length (Metric) | Particle Size | Price | Quantity & Availability | |||
71505014001
|
Thermo Scientific™
71505014001 |
4 mm | 10 mm | 5 μm |
Pack of 4 for $822.00
|
|
|||
Description
Thermo Scientific™ BetaBasic™ 18 Guard Cartridges are available in limited dimensions. Please contact Customer Service for more details.
- The 10mm design offers maximum protection with minimal increase in retention
- Fit Thermo Scientific UNIGUARD™ direct connection and stand alone holders
- Highly efficient and reproducible Thermo Scientific columns
- 150Å pore size for small molecules, peptides and protein digests
- Superb pH stability
Requires:
Thermo Scientific Unigard™ direct connection or stand-alone holder, sold separately.
Specifications
13% | |
10 mm | |
200 m2/g | |
Yes |
2 to 12 | |
150Å | |
Uniguard Direct-connection and Stand Alone Holder | |
L1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.