Learn More
New England Biolabs, Inc. beta-Nicotinamide adenine dinucleotide (NAD+) – 0.2 ml
Supplier: New England Biolabs, Inc. B9007S
Beta-Nicotinamide adenine dinucleotide (NAD+) is included with PreCR Repair Mix, an enzyme cocktail formulated to repair damaged template DNA prior to its use in the polymerase chain reaction (PCR), microarrays or other DNA technologies. NAD+ is supplied as an aqueous 50 mM solution.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.