Learn More
Siemens Healthineers Berichrom Heparin Reagent
Determination of heparin activity in plasma and for the monitoring of heparin therapy.
Supplier:  Siemens Healthineers 10446620
Catalog No. 23-044-703
This item is not returnable. View return policy
Description
Description
Berichrom Heparin, a chromogenic factor-Xa–based activity reagent, is used for the monitoring of heparin therapy and the determination of unfractionated (UF) and low-molecular-weight (LMW) heparin in patient samples. The reagent is not influenced by platelet factor IV complexing of heparin and offers a low detection limit of 0.05 IU/mL in UF heparin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Heparin Testing
3 x 1 mL
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.