Learn More
Electron Microscopy Sciences BenchRocker™ BenchRocker w/Flat Mat
Supplier:  Electron Microscopy Sciences 6570006
Encompass_Preferred
The BenchRocker 2D Rocker will provide the exact speeds and tilt angles needed for many molecular and biological mixing applications. Set the parameters to match your vessel size and volume of liquid for best results. Features a large 14 x 12" work surface and is safe for incubators and cold rooms.
Catalog No. 50-192-9977
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.