Learn More
Electron Microscopy Sciences BEEM Embedding Capsule Bottle Neck Tip
Supplier: Electron Microscopy Sciences 6991201
Specially shaped capsules are designed to overcome commonly encountered embedding problems; such as, small or thin elongated specimens and powder samples. These capsules are also good for centrifuging precipitated matters and allows for less trimming. BEEM Is A Registered Trademark of Better Equipment For Electron Microscopy, Inc. Bottle neck capsule produces a 7.9mm OD block with a hemihyperpoloid (rather than faced) tip.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.