Learn More
Electron Microscopy Sciences BEEM Dial-A-Grid & Block Storage
Supplier: Electron Microscopy Sciences 7114901
Two tone color coded plastic box with insert which has 24 letter-labeled crossed slots and 2 cavities for block storage. A rotating protection plate covers the slots and allows for exposure of one grid at a time. There are two additional cavities for block storage. BEEM Is A Registered Trademark of Better Equipment For Electron Microscopy, Inc.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.