Learn More
ILEX LIFE SCIENCES LLC BEADMATRIX PLUS FOR IPSCS 30G
SDP
Supplier:  ILEX LIFE SCIENCES LLC M0105X3
Encompass_Preferred
denovoMATRIX beadMATRIX+: precoated microcarriers for iPSCs culture, 30 g (3 x 10 g). DESCRIPTION: beadMATRIX+ are animal component-free, pre-coated microcarriers specifically designed for bioreactor-scale expansion of induced pluripotent stem cells (iPSCs) in serum-free and xeno-free media. The coating formulation is based on the successful 2D coating product myMATRIX iPSC and tailored to support customers in the establishment of translational cell therapies. Documentation describing quality and origin characteristics are provided. beadMATRIX+ is designed for the use in closed and controlled culture devices, e.g., stirred-tank bioreactors. ACCELERATION: enhance the development of your cell-based product. POTENCY: maintain stemness and genetic stability. ANIMAL-COMPONENT FREE: perform serum-free and xeno-free 3D-culture. Free of Endotoxins/BSE/TSE. COMPLIANT: developed in accordance with pharma grade requirements. BEAD MATERIAL: Cross-linked polystyrene. COATING: Biofunctional polymer.
Catalog No. 50-273-3908
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.