Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD BBL™ Prepared Plated Media: Phenylethyl Alcohol Agar with 5% Sheep Blood
For the isolation of gram-positive organisms, particularly gram-positive cocci, from specimens of mixed gram-positive and gram-negative flora and for determination of hemolytic reactions.
Supplier: BD 221179
This item is not returnable.
View return policy
Description
- Ready-to-use
- 100 x 10mm and 100 x 15mm-style plates
Specifications
ISO/DIS, 11014 | |
Mono-Plate Format | |
Phenylethyl Alcohol Agar with 5% Sheep Blood | |
Gram positive organisms, particularly gram positive cocci, from mixed specimens | |
20/Pk. |
Red | |
United States | |
Pancreatic digest of Casein 15g/L, Papaic digest of Soybean Meal 5g/L, Sodium Chloride 5g/L, beta-Phenylethyl Alcohol 2.5g/L, Agar 1.5g/L, Sheep Blood, defibrinated 5% | |
Plate |
Safety and Handling
ShelfLife : 84 Days
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction