Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Mycoflask™ TSI Agar™ Slants (Triple Sugar Iron Agar) Prepared Media

Used for the cultivation of fastidious and nonfastidious microorganisms from a variety of clinical and nonclinical specimens

$763.00

Specifications

Certifications/Compliance CE, AOAC, BAM, BS12, CCAM, CMPH2, COMPF, EP, ISO, MCM9, SMD, SMWW, USDA
Color Beige
Content And Storage 2°C to 8°C
Form Liquid
Manufacturing Origin United States
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BB21039
View Documents
BD
221039
100/Pk.
Pack of 100 for $763.00
Add to Cart
 
BB21038
View Documents
BD
221038
10/Pk. N/A N/A N/A
Description

Description

Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.

Specifications

Specifications

CE, AOAC, BAM, BS12, CCAM, CMPH2, COMPF, EP, ISO, MCM9, SMD, SMWW, USDA
2°C to 8°C
United States
Pancreatic digest of Casein 10g/L, Peptic digest of Animal Tissue 10g/L, Sodium Chloride 5g/L, Lactose 10g/L, Sucrose 10g/L, Dextrose 1g/L, Ferrous Ammonium Sulfate 0.2g/L, Sodium Thiosulfate 0.2g/L, Phenol red 0.025g/L, Agar 13g/L
Size K tubes with tight cap
Beige
Liquid
TSI Agar (Triple Sugar Iron)
Cultivation of fastidious and non-fastidious microorganisms from variety of clinical and nonclinical specimens
Videos
Safety and Handling

Safety and Handling

ShelfLife : 365 Days

SDS
Documents

Documents

Product Certifications

For in vitro diagnostic use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.