Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD BBL™ Mycoflask™ Mycosel™ Prepared Agar Slants
For the isolation of pathogenic fungi from materials having large amount of flora of other fungi and bacteria
Supplier: BD 220966
This item is not returnable.
View return policy
Description
- Selective medium containing cycloheximide and chloramphenicol
- For recovery of dermatophytes
Tubes, bottles and flasks with tight caps should be opened carefully to avoid injury due to breakage of glass.
Specifications
CE, BS12, CMPH2, MCM9 | |
2°C to 8°C | |
United States | |
Papaic digest of Soybean Meal 10g/L, Dextrose 10g/L, Agar 15.5g/L, Cycloheximide 0.4g/L, Chloramphenicol 0.05g/L | |
Tube |
Light Yellow | |
Liquid | |
Mycosel Agar | |
Isolation of pathogenic fungi from materials having large amount of flora of other fungi and bacteria | |
10/Pk. |
Safety and Handling
ShelfLife : 274 Days
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For in vitro Diagnostic Use
Spot an opportunity for improvement?Share a Content Correction