Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD BBL™ Prepared Plated Media: Columbia Agar with 5% Sheep Blood
For the isolation of microorganisms from samples or specimens
Supplier: BD 221165
This item is not returnable.
View return policy
Description
- Ready-to-use
- 100 x 10mm and 100 x 15mm-style plates
Specifications
ISO/DIS, 11014 | |
Solid | |
Columbia Agar with 5% Sheep Blood | |
Non-fastidious and fastidious microorganisms | |
20/Pk. |
Red | |
United States | |
Pancreatic digest of Casein 12g/L, Peptic digest of Animal Tissue 5g/L, Yeast extract 3.5g/L, Beef extract 3g/L, Corn Starch 1g/L, Sodium Chloride 5g/L, Agar 13.5g/L, Sheep Blood, Defibrinated 5% | |
Plate |
Safety and Handling
ShelfLife : 84 Days
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction