Learn More
BB700 Mouse Anti-Biotin, BD OptiBuild™
Mouse Monoclonal Antibody
Supplier:  BD Biosciences 758028
Catalog No. BDB758028
Explore all special offers
Add to Cart
Description
Description
Biotin Monoclonal antibody specifically detects Biotin. It is validated for Flow cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Biotin
Monoclonal
0.2 mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
Biotin-conjugated Keyhole Limpet Hemocyanin (KLH)
50 μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody.
IgG1 κ
Flow Cytometry
BK-1/39
Brilliant Blue 700 (BB700)
Mouse
Affinity Chromatography
RUO
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.