Learn More
Electron Microscopy Sciences NIGHTSEA™ Fluorescence Adapter Battery and Charger
Supplier: Electron Microscopy Sciences SFABATT
Compact battery pack that can run the Stereo Microscope Fluorescence Adapter (SFA) all day long. The battery enables truly portable operation so that you can set up demos anywhere or explore fluorescence in the field, without having to worry about plugging into the power grid.
Just plug the battery into the SFA base instead of the usual power connector. The battery has been tested at over 8 hours of continuous operation, and it will last even longer if you turn the system off when you don't need it. Plug it into the included charger overnight to recharge.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.