Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-Basophils, Alexa Fluor 647, Clone: 212H6.16, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals DDX0030A647100
Description
Basophils Monoclonal specifically detects Basophils in Human, Canine samples. It is validated for Flow Cytometry.Specifications
Basophils | |
Monoclonal | |
0.5mg/mL | |
Flow Cytometry 1:10-1:1000 | |
Anti-Basophils Basophil | |
Basophil was used as immunogen. | |
0.1 mg | |
Primary | |
Human, Canine | |
IgM |
Flow Cytometry | |
212H6.16 | |
Alexa Fluor 647 | |
NP_006857 | |
Mouse | |
Ion exchange chromatography | |
RUO | |
Human, Canine, No-cross reactivity on mouse basophil. | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction