Learn More
World Precision Instrument Barraquer Micro Needle Holder with Lock
Supplier: World Precision Instrument WP6036
Anis needle holders are surgical instruments designed for holding and manipulating needles during suturing procedures. These needle holders feature knurled round handles for a secure grip, facilitating precise control during surgery. The knurled texture enhances maneuverability, especially in delicate and fine tissue suturing. The 12 cm (4.7") version with a lock incorporates a mechanism that allows you to secure the needle in place, providing stability during the suturing process. This locking feature ensures that the needle remains firmly held, preventing unintentional slippage and contributing to the accuracy of the suturing. The 11.5 cm (4.5") version without a lock lack this mechanism, offering a more dynamic handling experience.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.