Learn More
NEOGEN Baird-Parker Agar, 500 g
SDP
Supplier:  NEOGEN 700003028
Encompass_Preferred
Baird-Parker Agar is used for detection and enumeration of Staphylococcus aureus in foods and is not intended for use in the diagnosis of disease or other conditions in humans.Baird-Parker Agar was first described in 1962. It is a selective medium for the isolation and presumptive identification of coagulase-positive staphylococci. This medium is used extensively for detecting Staphylococcus aureus in foods, dairy products, and other materials. Coagulase-positive staphylococci can grow and reproduce in cosmetic products. These products are tested for the presence of coagulase-positive staphylococci using standard microbiological methods.Formula:Enzymatic Digest of Casein - 10.0 g/LBeef Extract - 5.0 g/LYeast Extract - 1.0 g/LLithium Chloride - 5.0 g/LGlycine - 12.0 g/LSodium Pyruvate - 10.0 g/LAgar - 17.0 g/LFor more information visit https://www.neogen.com/solutions/Microbiology/baird-parker-agar
Catalog No. 50-201-5222
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.