Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
American Educational Products Bagged Specimen Collections

Each set contains six representative specimens.
Specifications
Grade | 5 to 12 |
---|---|
Includes | Resealable zip lock bag with a label |
Packaging | Specimen collections in bags |
Quantity | 6 Specimens |
Specimen Size | 3 to 3.8 cm |
Alert:
CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.
Includes: Identification key
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
S79094A
![]() |
American Educational Products
2221B |
Igneous Rock | N/A | N/A | N/A | ||||
S79096A
![]() |
American Educational Products
2231B |
Sedimentary Rock | N/A | N/A | N/A | ||||
S79095A
![]() |
American Educational Products
2226B |
Metamorphic Rock | N/A | N/A | N/A | ||||
Description
- Specimen size: 1.5 × 1.5 in. (3.8 × 3.8cm)
- Specimen names listed on bag
Specifications
5 to 12 | |
Specimen collections in bags | |
3 to 3.8 cm |
Resealable zip lock bag with a label | |
6 Specimens |
Spot an opportunity for improvement?
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.