Learn More
Andwin Scientific Reclosable Clear Bags, 6.3 x 7.6 cm, 1000/cs
Supplier: Andwin Scientific 89005300
Standard clear reclosable bags are available in 2 mm and 4 mm thickness. A wide zipper eases loading and provides a strong, secure seal. Bags 15.2 cm (6") in height or smaller are packed in perforated dispenser bags; all other bags are packed in inner bags without the dispenser bag.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.