Learn More
Andwin Scientific bright Yellow Biohazard Disposal Bag, 24/pk
SDP
Supplier:  Andwin Scientific 17911
Encompass_Preferred
Polyethylene Material Width 17 In. Length 30 In. Gauge 4 mil Hazardous Material Handle with Care.
Catalog No. NC1585509
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.