Learn More
Andwin Scientific Bacterial Cell Spreader, high impact polystyrene, pack of 100
SDP
Supplier:  Andwin Scientific 62452
Encompass_Preferred
The Bacterial Cell Spreader is constructed from high impact polystyrene and is ideal for easy spreading of cells onto the surface of an agar plate. Each sterilized spreader is individually packed to eliminate the risk of contamination. The spreader handle has an overall length of 176 millimeters and the spreader has an overall width of 36 millimeters. This product is sold in a pack of 100 green cell spreaders.
Catalog No. 50-727-642
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.