Learn More
Andwin Scientific BABCOCKMILK TST BOTTLEBRUSH10P
Supplier: Andwin Scientific 17050003
The Babcock Milk Test Bottle Brush is made to efficiently clean the narrow necks of Babcock milk test bottles. With short dense bristles, this product is able to easily reach into tall-form bottle necks. This product has natural white bristles that are twisted into a wire handle with hanging loop. It is sold in a pack of 10.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.