Learn More
STEMCELL Technologies Axygen AxyMats 96-Round-Well Sealing Mat for PCR Microplate, Size: 50 Units
SDP
Supplier:  STEMCELL Technologies 38107
Encompass_Preferred
Axygen AxyMats 96-Round-Well Sealing Mat for PCR Microplate is a silicone mat ideal for polymerase chain reaction (PCR) and storage of 96-round-well PCR microplates. The silicone mat minimizes evaporation in a wide range of temperatures. Made of research-grade silicone the sealing mat is autoclavable and can be washed with bleach and rinsed with ethanol for repeated use. Axygen AxyMats 96-Round-Well Sealing Mat for PCR Microplate is certified to be RNase/DNase-free and non-pyrogenic.
Catalog No. 50-239-0235
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.