Learn More
Enzo Life Sciences Axin-1 (human), (recombinant) (20µg)
SDP
Supplier:  Enzo Life Sciences ENZPRT1300020
Encompass_Preferred
Alternative name: Axis inhibition protein 1. Concentration: 0.05 µg/µl. Purity: ≥75%. Formulation: Liquid. In 50mM TRIS-HCl, pH 7.5, containing 50mM NaCl, 10mM gluthathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. Source: Produced in Sf9 insect cells. Produced in a baculovirus expression system. Long Term Storage: -80°C.
Catalog No. 50-201-3921
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.