Learn More
World Precision Instrument Autopsy Saw, Satterlee Bone Cutting, 30.5cm
Supplier: World Precision Instrument WP0129
The autopsy saw is for cutting bones in post-mortem exams. The end it rounded to reduce trauma to surrounding tissue. This saw is crafted from standard grade stainless steel, suitable for all forms of disinfection and autoclaving.