Learn More
Andwin Scientific AUTOPSY HANDLE FOR BLADE #4783
SDP
Supplier:  Andwin Scientific 4781
Encompass
This autopsy knife handle by Sakura Finetek is part of the Accu-Edge Semi-Disposable Autopsy Knife System. This specially designed plastic handle can hold all three sizes of blades that are made of high quality stainless steel. This product is autoclavable and is able to provide optimal balance and control to the user.
Catalog No. NC9449037
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.