Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Siemens Healthcare Diagnostics Atellica IM TSH3-Ultra II
An assay for in vitro diagnostic use in the quantitative determination of thyroid-stimulating hormone (TSH, thyrotropin) in human serum and plasma.
$654.00 - $3267.00
Specifications
CE Marker | Yes |
---|---|
Clia Complexity | Moderate Complexity |
Detectable Analytes | Thyroid Stimulating Hormone (TSH) |
Detection Method | Chemiluminescence |
DoA Calibrators | Yes |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
21-279-173
|
Siemens Healthcare Diagnostics
11208706 |
130 Tests |
Pack of 130 for $654.00
|
|
|||||
21-279-172
|
Siemens Healthcare Diagnostics
11208707 |
650 Tests |
Pack of 650 for $3,267.00
|
|
|||||
Description
An assay for in vitro diagnostic use in the quantitative determination of thyroid-stimulating hormone (TSH, thyrotropin) in human serum and plasma.Specifications
Yes | |
Thyroid Stimulating Hormone (TSH) | |
Yes | |
Assay | |
BSA, BGG, goat serum, sodium azide (<0.1%), surfactant, preservatives | |
Assay is linear for the measuring interval of 0.010 to 150.000 μL U/mL (mL U/L). Linearity testing was performed in accordance with CLSI Document EP06-ed2.22. | |
Liquid Ready-to-Use |
Moderate Complexity | |
Chemiluminescence | |
Serum, EDTA plasma, Heparinized plasma | |
Single-use | |
Atellica IM Analyzer | |
14 Minutes |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title