Learn More
Hach Company Total Chlorine Chemkey Reagents
Supplier:  Hach Company 9429100
Encompass_Preferred
Total Chlorine Chemkeys, range 0.04 - 10 mg/L Cl2. For SL1000 Portable Parallel Analyzer (PPA). Box of 25.USEPA accepted for reporting in drinking water analysis using Hach Method 10260.
Catalog No. 50-199-7338
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.