Learn More
Hach Company High Range Hardness Chemkey Reagents (box of 25)
Supplier:  Hach Company 8636300
Encompass_Preferred
Avoid the hassle by avoiding titrations with our easy to use colorimetric Hardness Chemkey. The Hach SL1000 Portable Parallel Analyzer (PPA) performs the same tests with less than half the manual steps. Get highly accurate results, with less opportunity for errors, in a fraction of the time. Up to six parameters, tested in parallel. The SL1000 is a portable colorimeter that provides faster testing by using Chemkey reagents instead of powder pillows. All chemicals and processes are entirely contained inside the Chemkey.High Range Hardness Chemkeys, range 90 - 750 mg/L CaCO3. For SL1000 Portable Parallel Analyzer (PPA).
Catalog No. 50-199-7274
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.