Learn More
Eurofins Discoverx AssayComplete Thawing Reagent T1, 100. (92-4101TR)
Supplier: Eurofins Discoverx 924101TR
AssayComplete Thawing Reagent T1 is an optimized thawing reagent for revival and recovery of Eurofins DiscoverX cell lines. This reagent ensures optimal post-thaw viability, recovery and assay performance upon thawing. This product is provided as a bottle of 100 mL. Please refer to cell line specific datasheet to determine the recommended thawing reagent. For detailed product information, User manual and datasheet, visit www.discoverx.com/reagents.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.