Learn More
Enzo Life Sciences Argatroban (10mg). CAS: 74863-84-6
SDP
Supplier:  Enzo Life Sciences BMLPI1460010
Encompass_Preferred
Argatroban is a synthetic peptidomimetic thrombin inhibitor which is in clinical use. It presents significant advantages as an anticoagulant in deep vein thrombosis, stroke prevention in atrial fibrillation and thromboembolic disease and especially in patients with coronary artery disease and heparin-induced thrombocytopenia. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (25mg/ml) or 100% ethanol (25mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-1487
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.