Learn More
Andwin Scientific AQUAMOUNT
Supplier: Andwin Scientific 41799008
The Aqua-Mount Mounting Medium is a non-fluorescing, aqueous formula that is ideal for immunostaining procedures. This product is made from well and strictly regulated procedures to guarantee the product's consistency. Every item has gone tissue testings in order to validate its effectivity. This medium is sold individually in a quantity of 20 mL.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.