Learn More
Andwin Scientific AQUAMONT M800 20ML
SDP
Supplier:  Andwin Scientific M800
Encompass_Preferred
Aqua-Mount Mounting Medium is a non-fluorescing, aqueous formula that is made to be utilized for immunostaining. It is sold in a quantity of 20 mL.
Catalog No. NC9821477
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.