Learn More
New England Biolabs, Inc. Apyrase – 10 units
SDP
Supplier:  New England Biolabs, Inc. M0398S
Encompass
Apyrase (recombinant, E. coli) is a highly active ATP-diphosphohydrolase that catalyses the sequential hydrolysis of ATP to ADP and ADP to AMP releasing inorganic phosphate. It is a recombinant version of one of several isoforms of apyrase. It can also hydrolyse 5' tri- and diphosphate ribonucleosides and deoxyribonucleosides to their respective 5' monophosphates. Apyrase can catalyse the conversion of 5' triphosphorylated RNA to 5' monophosphorylated RNA.
Catalog No. 50-104-7495
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.